siRNA Details
| Gene Symbol: Crk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277219.1 | NP_001264148.1 | 3 | 798 | ||
| NM_001277221.1 | NP_001264150.1 | 2 | 432 | |||
| NM_133656.5 | NP_598417.2 | 3 | 968 | |||
| GenBank mRNA | AK028179.1 | - | 3 | 775 | ||
| AK028488.1 | - | 3 | 918 | |||
| AK036905.1 | - | 3 | 749 | |||
| AK052795.1 | - | 3 | 757 | |||
| AK076097.1 | - | 3 | 663 | |||
| AK088519.1 | - | 3 | 769 | |||
| AK139089.1 | - | 3 | 754 | |||
| AK152549.1 | - | 3 | 942 | |||
| AK154987.1 | - | 3 | 953 | |||
| AK161761.1 | - | 3 | 910 | |||
| AK163291.1 | - | 2 | 407 | |||
| AK198502.1 | - | 2 | 117 | |||
| AK200050.1 | - | 2 | 93 | |||
| AK202550.1 | - | 2 | 53 | |||
| AK212188.1 | - | 2 | 104 | |||
| BC012216.1 | - | 3 | 904 | 3992341 | ||
| BC031149.1 | - | 3 | 731 | 4975654 | ||
| S72408.1 | - | 3 | 893 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_019302, XM_006246913, D44481, FQ210506.

Crk
Genomic Map