siRNA Details
| Gene Symbol: Ier3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133662.2 | NP_598423.1 | 2 | 248 | ||
| GenBank mRNA | AK003466.1 | - | 2 | 274 | ||
| AK051003.1 | - | 2 | 278 | |||
| AK170477.1 | - | 2 | 278 | |||
| BC006950.1 | - | 2 | 248 | 2811734 | ||
| CT010247.1 | - | 2 | 242 | |||
| CT010271.1 | - | 2 | 242 | |||
| X67644.1 | - | 1062 | ||||

Ier3
Genomic Map