siRNA Details
| Gene Symbol: Sac3d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133678.3 | NP_598439.3 | 2 | 965 | ||
| GenBank mRNA | AJ131957.2 | - | 2 | 959 | ||
| AK011298.1 | - | 2 | 877 | |||
| AK014138.1 | - | 2 | 919 | |||
| AK075930.1 | - | 2 | 925 | |||
| AK139556.1 | - | 2 | 925 | |||
| BC144812.1 | - | 2 | 1029 | 9053360 | ||
| BC145789.1 | - | 2 | 1029 | 40131092 | ||

Sac3d1
Genomic Map