siRNA Details
| Gene Symbol: Rbm42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133693.2 | NP_598454.2 | 7 | 891 | ||
| GenBank mRNA | AK005626.1 | - | 4 | 328 | ||
| AK013929.1 | - | 7 | 832 | |||
| AK151000.1 | - | 7 | 803 | |||
| AK217622.1 | - | 4 | 345 | |||
| BC009148.1 | - | 7 | 828 | 3600406 | ||
| BC011286.1 | - | 7 | 810 | 3485410 | ||
| BC027372.1 | - | 7 | 815 | 4952304 | ||
| BC039917.1 | - | 7 | 826 | |||
| BC057928.1 | - | 7 | 817 | 5289543 | ||

Rbm42
Genomic Map