siRNA Details
| Gene Symbol: Prss22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133731.2 | NP_598492.2 | 4 | 454 | ||
| GenBank mRNA | AB010778.1 | - | 4 | 437 | ||
| AK014645.1 | - | 4 | 454 | |||
| BC055854.1 | - | 4 | 439 | 4035351 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss22| Gene Symbol: Prss22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133731.2 | NP_598492.2 | 4 | 454 | ||
| GenBank mRNA | AB010778.1 | - | 4 | 437 | ||
| AK014645.1 | - | 4 | 454 | |||
| BC055854.1 | - | 4 | 439 | 4035351 | ||