siRNA Details
| Gene Symbol: Msl3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163833.1 | NP_001157305.1 | 2 | 539 | ||
| GenBank mRNA | AK155382.1 | - | 2 | 525 | ||
| BC013191.1 | - | 1 | 141 | |||
| BC037487.1 | - | 2 | 550 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msl3l2| Gene Symbol: Msl3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163833.1 | NP_001157305.1 | 2 | 539 | ||
| GenBank mRNA | AK155382.1 | - | 2 | 525 | ||
| BC013191.1 | - | 1 | 141 | |||
| BC037487.1 | - | 2 | 550 | |||