siRNA Details
| Gene Symbol: Tor1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133673.3 | NP_598434.2 | 2 | 313 | ||
| GenBank mRNA | AJ297743.1 | - | 2 | 343 | ||
| AK017290.1 | - | 2 | 332 | |||
| AK028859.1 | - | 2 | 127 | |||
| AK054404.1 | - | 2 | 344 | |||
| AK149754.1 | - | 2 | 334 | |||
| AK159843.1 | - | 2 | 312 | |||
| AK169692.1 | - | 2 | 336 | |||
| BC018456.1 | - | 2 | 312 | 3582246 | ||

Tor1b
Genomic Map