siRNA Details
| Gene Symbol: Rab31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133685.2 | NP_598446.2 | 4, 5 | 489 | ||
| GenBank mRNA | AB232617.1 | - | 4, 5 | 281 | ||
| AK051270.1 | - | 4, 5 | 409 | |||
| AK156677.1 | - | 4, 5 | 490 | |||
| AK159095.1 | - | 4, 5 | 378 | |||
| BC013063.1 | - | 4, 5 | 418 | 3497938 | ||

Rab31
Genomic Map