siRNA Details
| Gene Symbol: Qprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133686.1 | NP_598447.1 | 3 | 643 | ||
| GenBank mRNA | AK010602.1 | - | 3 | 656 | ||
| BC011191.1 | - | 3 | 643 | 4196244 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Qprt| Gene Symbol: Qprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133686.1 | NP_598447.1 | 3 | 643 | ||
| GenBank mRNA | AK010602.1 | - | 3 | 656 | ||
| BC011191.1 | - | 3 | 643 | 4196244 | ||