siRNA Details
| Gene Symbol: Cxxc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133687.2 | NP_598448.1 | 2 | 426 | ||
| GenBank mRNA | AK005364.1 | - | 2 | 455 | ||
| AK015150.1 | - | 2 | 330 | |||
| AK132950.1 | - | 2 | 350 | |||
| BC016207.1 | - | 1 | 117 | 4912395 | ||
| BC089314.1 | - | 2 | 426 | 30541251 | ||

Cxxc5
Genomic Map