siRNA Details
| Gene Symbol: Asnsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133728.3 | NP_598489.2 | 4 | 625 | ||
| GenBank mRNA | AK008875.1 | - | 4 | 542 | ||
| AK031295.1 | - | 4 | 583 | |||
| AK034746.1 | - | 544 | ||||
| AK049194.1 | - | 4 | 572 | |||
| AK050045.1 | - | 4 | 564 | |||
| AK076793.1 | - | 4 | 527 | |||
| AK154984.1 | - | 5 | 722 | |||
| AK155030.1 | - | 4 | 564 | |||
| AK168103.1 | - | 4 | 565 | |||
| AK170510.1 | - | 4 | 541 | |||
| BC013617.1 | - | 4 | 445 | 3979452 | ||

Asnsd1
Genomic Map