siRNA Details
| Gene Symbol: Fam118a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133750.4 | NP_598511.1 | 3 | 436 | ||
| GenBank mRNA | AK078807.1 | - | 1 | 174 | ||
| AK082151.1 | - | 3 | 374 | |||
| AK143690.1 | - | 3 | 409 | |||
| BC016425.1 | - | 3 | 294 | 4487028 | ||
| BC058794.1 | - | 3 | 497 | 6486049 | ||
| BC066174.1 | - | 3 | 352 | 30109713 | ||

Fam118a
Genomic Map