siRNA Details
| Gene Symbol: Lad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133664.3 | NP_598425.2 | 10 | 2595 | ||
| GenBank mRNA | AK083526.1 | - | 10 | 2596 | ||
| BC016257.1 | - | 10 | 2266 | 4912735 | ||
| BC025580.1 | - | 10 | 2499 | 5325039 | ||
| BC031131.1 | - | 10 | 2442 | 4954891 | ||

Lad1
Genomic Map