siRNA Details
| Gene Symbol: Tmem19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133683.3 | NP_598444.1 | 7 | 1886 | ||
| GenBank mRNA | AK034298.1 | - | 7 | 1892 | ||
| AK036228.1 | - | 6 | 1963 | |||
| AK042426.1 | - | 6 | 1941 | |||
| AK136778.1 | - | 1876 | ||||
| AK138931.1 | - | 7 | 1874 | |||
| AK155275.1 | - | 7 | 1847 | |||
| AK170143.1 | - | 7 | 1846 | |||
| AK171087.1 | - | 7 | 1846 | |||
| AK189585.1 | - | 1 | 78 | |||
| BC016895.1 | - | 7 | 1888 | 4239007 | ||

Tmem19
Genomic Map