siRNA Details
| Gene Symbol: Rab31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133685.2 | NP_598446.2 | 7 | 1240 | ||
| GenBank mRNA | AK007048.1 | - | 3 | 981 | ||
| AK016608.1 | - | 3 | 969 | |||
| AK051270.1 | - | 7 | 1160 | |||
| AK156677.1 | - | 7 | 1243 | |||
| AK159095.1 | - | 7 | 1131 | |||
| BC013063.1 | - | 7 | 1169 | 3497938 | ||

Rab31
Genomic Map