siRNA Details
| Gene Symbol: Rbm42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133693.2 | NP_598454.2 | 10 | 1569 | ||
| GenBank mRNA | AK013929.1 | - | 10 | 1510 | ||
| AK151000.1 | - | 10 | 1481 | |||
| AK208148.1 | - | 1 | 41 | |||
| AK211115.1 | - | 1 | 41 | |||
| BC009148.1 | - | 10 | 1506 | 3600406 | ||
| BC011286.1 | - | 10 | 1488 | 3485410 | ||
| BC027372.1 | - | 10 | 1493 | 4952304 | ||
| BC039917.1 | - | 10 | 1504 | |||
| BC057928.1 | - | 10 | 1495 | 5289543 | ||
| BY551742.1 | - | 3 | 369 | |||

Rbm42
Genomic Map