siRNA Details
| Gene Symbol: Ctdspl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133710.3 | NP_598471.3 | 8 | 1948 | ||
| GenBank mRNA | AJ344340.1 | - | 8 | 2030 | ||
| AK138484.1 | - | 1 | 678 | |||
| AK171161.1 | - | 1 | 678 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctdspl| Gene Symbol: Ctdspl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133710.3 | NP_598471.3 | 8 | 1948 | ||
| GenBank mRNA | AJ344340.1 | - | 8 | 2030 | ||
| AK138484.1 | - | 1 | 678 | |||
| AK171161.1 | - | 1 | 678 | |||