siRNA Details
| Gene Symbol: Tmem30a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133718.4 | NP_598479.1 | 7 | 1712 | ||
| GenBank mRNA | AK008408.1 | - | 7 | 1597 | ||
| AK077065.1 | - | 7 | 1715 | |||
| AK150071.1 | - | 7 | 1615 | |||
| AK159364.1 | - | 7 | 1639 | |||
| AK170171.1 | - | 7 | 1615 | |||
| AK170697.1 | - | 7 | 1616 | |||
| BC018367.1 | - | 7 | 1564 | 2648510 | ||
| BC018491.1 | - | 7 | 1592 | 2651602 | ||
| BC026136.1 | - | 6 | 1188 | |||

Tmem30a
Genomic Map