siRNA Details
| Gene Symbol: Kptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133727.2 | NP_598488.2 | 12 | 1611 | ||
| GenBank mRNA | AK009746.1 | - | 12 | 1591 | ||
| BC018316.1 | - | 12 | 1579 | 4160465 | ||
| BU953076.1 | - | 2 | 524 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kptn| Gene Symbol: Kptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133727.2 | NP_598488.2 | 12 | 1611 | ||
| GenBank mRNA | AK009746.1 | - | 12 | 1591 | ||
| BC018316.1 | - | 12 | 1579 | 4160465 | ||
| BU953076.1 | - | 2 | 524 | |||