siRNA Details
| Gene Symbol: Opa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199177.1 | NP_001186106.1 | 19, 20 | 2037 | ||
| NM_133752.3 | NP_598513.1 | 18, 19 | 1983 | |||
| GenBank mRNA | AB044138.1 | - | 18, 19 | 1975 | ||
| AK004715.1 | - | 19, 20 | 1993 | |||
| AK029157.1 | - | 18, 19 | 1941 | |||
| AK030835.1 | - | 19, 20 | 1991 | |||
| AK122311.1 | - | 18, 19 | 1912 | |||
| AK145620.1 | - | 18, 19 | 1935 | |||
| BC052704.1 | - | 17, 18 | 2775 | |||
| BC138665.1 | - | 18, 19 | 1864 | 8861686 | ||
| BC145959.1 | - | 18, 19 | 1864 | 40131262 | ||

Opa1
Genomic Map