siRNA Details
| Gene Symbol: F12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000505.3 | NP_000496.2 | 5 | 376 | ||
| XM_011534462.2 | XP_011532764.1 | 2 | 210 | |||
| GenBank mRNA | AB095845.1 | - | 5 | 376 | ||
| M11723.1 | - | 5 | 288 | |||
| M31315.1 | - | 5 | 318 | |||

F12
Genomic Map