siRNA Details
| Gene Symbol: Nol12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133800.3 | NP_598561.2 | 2 | 182 | ||
| GenBank mRNA | AK036891.1 | - | 2 | 125 | ||
| AK037035.1 | - | 2 | 176 | |||
| AK083539.1 | - | 184 | ||||
| AK167446.1 | - | 2 | 176 | |||
| AY940114.1 | - | 2 | 125 | |||
| BC094424.1 | - | 2 | 169 | 6819926 | ||

Nol12
Genomic Map