siRNA Details
| Gene Symbol: Hnrnpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166427.1 | NP_001159899.1 | 3 | 498 | ||
| NM_001166428.1 | NP_001159900.1 | 4 | 437 | |||
| NM_001166429.1 | NP_001159901.1 | 4 | 398 | |||
| NM_001166430.1 | NP_001159902.1 | 4 | 749 | |||
| NM_001166431.1 | NP_001159903.1 | 3 | 688 | |||
| NM_001166432.1 | NP_001159904.1 | 4 | 489 | |||
| NM_133834.2 | NP_598595.1 | 4 | 380 | |||
| GenBank mRNA | AK076478.1 | - | 3 | 338 | ||
| AU079872.1 | - | 4 | 563 | |||
| BC018185.1 | - | 3 | 323 | 3995067 | ||
| BC023793.1 | - | 4 | 375 | |||
| BC025481.1 | - | 4 | 357 | 5354000 | ||
| BC029163.1 | - | 1 | 141 | 4949890 | ||
| BC032197.1 | - | 4 | 376 | |||
| BC033483.1 | - | 1 | 139 | 4950131 | ||
| BC042689.1 | - | 3 | 621 | |||
| BC047346.1 | - | 3 | 621 | |||
| BC089313.1 | - | 4 | 222 | 6808998 | ||
| CA539382.1 | - | 4 | 377 | |||
| CF163622.1 | - | 4 | 505 | |||
| CJ048596.1 | - | 4 | 383 | |||
| CJ054614.1 | - | 4 | 416 | |||
| CK389060.1 | - | 4 | 389 | |||

Hnrnpf
Genomic Map