siRNA Details
| Gene Symbol: HBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000518.4 | NP_000509.1 | 3 | 584 | ||
| GenBank mRNA | AF181832.1 | - | 3 | 477 | ||
| AF181989.1 | - | 3 | 567 | |||
| AF349114.1 | - | 3 | 569 | |||
| AY509193.1 | - | 3 | 584 | |||
| M11428.1 | - | 1 | 171 | |||
| M25113.1 | - | 3 | 534 | |||
| V00497.1 | - | 3 | 584 | |||
| V00500.1 | - | 3 | 534 | |||

HBB
Genomic Map