siRNA Details
Gene Symbol: Ube3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133907.3 | NP_598668.1 | 1, 2 | 347 | ||
GenBank mRNA | AK029973.1 | - | 1, 2 | 157 | ||
AK045211.1 | - | 1, 2 | 144 | |||
AK046056.1 | - | 1, 2 | 348 | |||
AK122187.1 | - | 1, 2 | 282 | |||
AK140491.1 | - | 1, 2 | 104 | |||
BC059245.1 | - | 1, 2 | 163 | |||
BC120731.1 | - | 1, 2 | 94 | 40129654 | ||
BC137626.1 | - | 1, 2 | 94 | 8860645 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
