siRNA Details
| Gene Symbol: Itfg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133927.1 | NP_598688.1 | 2, 3 | 291 | ||
| GenBank mRNA | AK134630.1 | - | 2, 3 | 312 | ||
| AK147728.1 | - | 2, 3 | 293 | |||
| AK159637.1 | - | 2, 3 | 300 | |||
| AK183090.1 | - | 1, 2 | 36 | |||
| BC014833.1 | - | 2, 3 | 291 | 4507661 | ||
| BC025492.1 | - | 2, 3 | 311 | 5309950 | ||

Itfg2
Genomic Map