siRNA Details
Gene Symbol: Adgrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133776.2 | NP_598537.2 | 5 | 453 | ||
GenBank mRNA | AK019869.1 | - | 5 | 456 | ||
BC019217.1 | - | 5 | 482 | 5065702 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adgrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133776.2 | NP_598537.2 | 5 | 453 | ||
GenBank mRNA | AK019869.1 | - | 5 | 456 | ||
BC019217.1 | - | 5 | 482 | 5065702 |