siRNA Details
| Gene Symbol: Stk17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133810.3 | NP_598571.2 | 3 | 485 | ||
| GenBank mRNA | AK029551.1 | - | 3 | 426 | ||
| AK035693.1 | - | 3 | 453 | |||
| AK076932.1 | - | 3 | 452 | |||
| AK083332.1 | - | 3 | 454 | |||
| AK153108.1 | - | 3 | 429 | |||
| AK165291.1 | - | 3 | 425 | |||
| AY092028.1 | - | 3 | 438 | |||
| BC006579.1 | - | 3 | 440 | 3586767 | ||

Stk17b
Genomic Map