siRNA Details
| Gene Symbol: Tm9sf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133847.3 | NP_598608.2 | 4 | 537 | ||
| GenBank mRNA | AB093220.2 | - | 3 | 281 | ||
| AK035622.1 | - | 4 | 499 | |||
| AK047497.1 | - | 4 | 291 | |||
| AK048158.1 | - | 4 | 295 | |||
| AK083541.1 | - | 4 | 537 | |||
| AK087048.1 | - | 4 | 290 | |||
| AK088012.1 | - | 4 | 293 | |||
| AK162435.1 | - | 4 | 298 | |||
| AK171487.1 | - | 4 | 295 | |||
| BC110309.1 | - | 4 | 293 | 5324218 | ||

Tm9sf4
Genomic Map