siRNA Details
| Gene Symbol: Nusap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042652.1 | NP_001036117.1 | 2, 3 | 675 | ||
| NM_133851.3 | NP_598612.1 | 2, 3 | 675 | |||
| GenBank mRNA | AF305710.1 | - | 2, 3 | 256 | ||
| AK042697.1 | - | 256 | ||||
| AK044819.1 | - | 2, 3 | 248 | |||
| AK076028.1 | - | 2, 3 | 245 | |||
| AK088389.1 | - | 2, 3 | 245 | |||
| BC004787.1 | - | 2, 3 | 225 | 3588697 | ||
| BC100392.1 | - | 2, 3 | 675 | 30110191 | ||

Nusap1
Genomic Map