siRNA Details
| Gene Symbol: Olfml3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133859.2 | NP_598620.2 | 2 | 349 | ||
| GenBank mRNA | AK002518.1 | - | 2 | 313 | ||
| AK076084.1 | - | 2 | 315 | |||
| AK131879.1 | - | 313 | ||||
| AK143728.1 | - | 901 | ||||
| BC005485.1 | - | 2 | 296 | 3485412 | ||
| BY059553.1 | - | 2 | 350 | |||

Olfml3
Genomic Map