siRNA Details
| Gene Symbol: Cept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293694.1 | NP_001280623.1 | 3 | 623 | ||
| NM_133869.4 | NP_598630.2 | 3 | 623 | |||
| GenBank mRNA | AK037116.1 | - | 3 | 583 | ||
| BC021753.1 | - | 3 | 552 | 4167222 | ||
| BC023783.1 | - | 3 | 563 | 5346896 | ||

Cept1
Genomic Map