siRNA Details
| Gene Symbol: Zbtb48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133879.2 | NP_598640.2 | 2 | 560 | ||
| GenBank mRNA | AK027912.1 | - | 2 | 561 | ||
| AM161454.1 | - | 2 | 508 | |||
| BC005515.1 | - | 2 | 571 | 3493611 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb48| Gene Symbol: Zbtb48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133879.2 | NP_598640.2 | 2 | 560 | ||
| GenBank mRNA | AK027912.1 | - | 2 | 561 | ||
| AM161454.1 | - | 2 | 508 | |||
| BC005515.1 | - | 2 | 571 | 3493611 | ||