siRNA Details
Gene Symbol: Gpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290742.1 | NP_001277671.1 | 4 | 784 | ||
NM_133884.2 | NP_598645.2 | 4 | 853 | |||
GenBank mRNA | BC018407.1 | - | 4 | 846 | 4015340 | |
CN721622.1 | - | 4 | 455 |
