siRNA Details
| Gene Symbol: Slc44a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133891.3 | NP_598652.3 | 16 | 2231 | ||
| GenBank mRNA | AY249865.1 | - | 16 | 1987 | ||
| BC010258.1 | - | 4 | 425 | |||
| BC025941.1 | - | 7 | 793 | |||
| BC113167.1 | - | 16 | 1987 | 40059854 | ||
| BC113169.1 | - | 16 | 1987 | 40059860 | ||

Slc44a1
Genomic Map