siRNA Details
| Gene Symbol: SFTPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000542.3 | NP_000533.3 | 4, 5 | 396 | ||
| NM_198843.2 | NP_942140.2 | 4, 5 | 396 | |||
| XM_005264487.2 | XP_005264544.1 | 3, 4 | 311 | |||
| XM_005264488.3 | XP_005264545.2 | 3, 4 | 276 | |||
| XM_005264490.4 | XP_005264547.2 | 3, 4 | 276 | |||
| XM_017004702.1 | XP_016860191.1 | 3, 4 | 311 | |||
| XM_017004703.1 | XP_016860192.1 | 3, 4 | 276 | |||
| GenBank mRNA | AL548140.3 | - | 4, 5 | 396 | ||
| BC032785.1 | - | 3, 4 | 263 | 5179408 | ||
| BI821350.1 | - | 266 | ||||
| DQ317589.1 | - | 3, 4 | 249 | |||
| J02761.1 | - | 3, 4 | 273 | |||

SFTPB
Genomic Map