siRNA Details
| Gene Symbol: Stard4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133774.4 | NP_598535.1 | 6 | 2483 | ||
| GenBank mRNA | AK014587.1 | - | 1 | 2278 | ||
| AK048754.1 | - | 1 | 522 | |||
| AK054501.1 | - | 6 | 2484 | |||
| AK085331.1 | - | 6 | 2487 | |||

Stard4
Genomic Map