siRNA Details
| Gene Symbol: Adgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133776.2 | NP_598537.2 | 16 | 3144 | ||
| GenBank mRNA | AK052799.1 | - | 5 | 815 | ||
| AW113731.1 | - | 1 | 148 | |||
| BB506964.2 | - | 3 | 344 | |||
| BC019217.1 | - | 16 | 3173 | 5065702 | ||

Adgrf1
Genomic Map