siRNA Details
| Gene Symbol: Ubac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133835.2 | NP_598596.2 | 7 | 1094 | ||
| GenBank mRNA | AK004060.1 | - | 1 | 182 | ||
| AK157857.1 | - | 7 | 928 | |||
| AK161969.1 | - | 2 | 297 | |||
| AK177482.1 | - | 2 | 278 | |||
| AK209158.1 | - | 2 | 270 | |||
| AK209934.1 | - | 1 | 46 | |||
| AK213285.1 | - | 2 | 267 | |||
| BC021811.1 | - | 7 | 1045 | 3662474 | ||

Ubac1
Genomic Map