siRNA Details
| Gene Symbol: Tm9sf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133847.3 | NP_598608.2 | 18 | 2472 | ||
| GenBank mRNA | AB093220.2 | - | 17 | 2216 | ||
| AK048158.1 | - | 18 | 2230 | |||
| AK083541.1 | - | 18 | 2472 | |||
| AK087048.1 | - | 18 | 2226 | |||
| AK088012.1 | - | 18 | 2229 | |||
| AK162435.1 | - | 18 | 2233 | |||
| BC006741.1 | - | 4 | 909 | |||
| BC071208.1 | - | 5 | 1291 | |||
| BC110309.1 | - | 18 | 2228 | 5324218 | ||
| BC116320.2 | - | 18 | 2424 | 40096002 | ||

Tm9sf4
Genomic Map