siRNA Details
| Gene Symbol: Hrh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133849.3 | NP_598610.1 | 3 | 2196 | ||
| GenBank mRNA | AK140374.1 | - | 3 | 2201 | ||
| AK156130.1 | - | 1 | 1523 | |||
| AY044153.2 | - | 3 | 2023 | |||
| BC028947.1 | - | 1 | 1346 | |||
More Information
Set Membership: |
|

Hrh3
Genomic Map