siRNA Details
| Gene Symbol: Dclre1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025312.1 | NP_001020483.1 | 3 | 1218 | ||
| NM_133865.2 | NP_598626.2 | 4 | 1384 | |||
| GenBank mRNA | AK049115.1 | - | 4 | 1379 | ||
| AK051104.1 | - | 4 | 1339 | |||
| AK084347.1 | - | 3 | 1213 | |||
| AK134324.1 | - | 4 | 1144 | |||
| AK137495.1 | - | 4 | 941 | |||
| AK138568.1 | - | 4 | 1442 | |||
| AK154124.1 | - | 4 | 1286 | |||
| BC011094.1 | - | 3 | 843 | 3493038 | ||
| BC067017.1 | - | 3 | 1127 | 6827667 | ||
| BC125277.1 | - | 3 | 839 | |||
| BC144725.1 | - | 3 | 829 | |||
| CA317555.1 | - | 1 | 103 | |||

Dclre1b
Genomic Map