siRNA Details
| Gene Symbol: Gpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290742.1 | NP_001277671.1 | 5 | 1059 | ||
| NM_133884.2 | NP_598645.2 | 5 | 1128 | |||
| GenBank mRNA | BC018407.1 | - | 5 | 1121 | 4015340 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpn2| Gene Symbol: Gpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290742.1 | NP_001277671.1 | 5 | 1059 | ||
| NM_133884.2 | NP_598645.2 | 5 | 1128 | |||
| GenBank mRNA | BC018407.1 | - | 5 | 1121 | 4015340 | |