siRNA Details
| Gene Symbol: Ube3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133907.3 | NP_598668.1 | 4 | 577 | ||
| GenBank mRNA | AK029973.1 | - | 4 | 387 | ||
| AK045211.1 | - | 4 | 374 | |||
| AK046056.1 | - | 4 | 578 | |||
| AK122187.1 | - | 4 | 512 | |||
| AK140491.1 | - | 4 | 334 | |||
| BC059245.1 | - | 4 | 392 | |||
| BC120731.1 | - | 4 | 324 | 40129654 | ||
| BC137626.1 | - | 4 | 324 | 8860645 | ||

Ube3c
Genomic Map