siRNA Details
| Gene Symbol: Ttll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142732.1 | NP_001136204.1 | 5 | 731 | ||
| NM_133923.6 | NP_598684.4 | 5 | 848 | |||
| GenBank mRNA | AK029462.1 | - | 5 | 732 | ||
| AK080321.1 | - | 1 | 262 | |||
| AM690746.1 | - | 5 | 848 | |||
| BC006830.1 | - | 3 | 276 | |||
| BC021404.1 | - | 4 | 480 | |||
| CF916454.1 | - | 1 | 97 | |||

Ttll3
Genomic Map