siRNA Details
| Gene Symbol: Brk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133937.1 | NP_598698.1 | 3 | 225 | ||
| GenBank mRNA | AK003342.1 | - | 3 | 240 | ||
| AK020128.1 | - | 3 | 240 | |||
| AK020277.1 | - | 3 | 241 | |||
| AK150689.1 | - | 3 | 240 | |||
| AK163236.1 | - | 3 | 240 | |||
| AK190298.1 | - | 3 | 164 | |||
| AK193342.1 | - | 3 | 164 | |||
| BC010582.1 | - | 3 | 225 | 3154777 | ||

Brk1
Genomic Map