siRNA Details
| Gene Symbol: Numa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133947.3 | NP_598708.3 | 7 | 578 | ||
| GenBank mRNA | AK032693.1 | - | 7 | 627 | ||
| AK050670.1 | - | 7 | 659 | |||
| AK168399.1 | - | 7 | 596 | |||
| BC049791.1 | - | 7 | 578 | 6505037 | ||

Numa1
Genomic Map