siRNA Details
| Gene Symbol: Tbrg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130457.1 | NP_001123929.1 | 3 | 416 | ||
| NM_134011.2 | NP_598772.1 | 3 | 416 | |||
| GenBank mRNA | AK028966.1 | - | 3 | 394 | ||
| AK129245.1 | - | 3 | 371 | |||
| AK144813.1 | - | 3 | 414 | |||
| AK166760.1 | - | 3 | 414 | |||
| AK168088.1 | - | 3 | 414 | |||
| AK188017.1 | - | 1 | 31 | |||
| BC016483.1 | - | 3 | 372 | 3155271 | ||
| BC031155.1 | - | 3 | 387 | 4976690 | ||
| BC031910.1 | - | 3 | 386 | 5362943 | ||

Tbrg4
Genomic Map