siRNA Details
Gene Symbol: Mat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199274.1 | NP_001186203.1 | 1, 2 | 412 | ||
NM_134017.2 | NP_598778.1 | 1, 2 | 216 | |||
GenBank mRNA | AB070267.1 | - | 1, 2 | 183 | ||
AK038303.1 | - | 1, 2 | 163 | |||
AK075600.1 | - | 1, 2 | 117 | |||
AK075947.1 | - | 1, 2 | 151 | |||
AK153126.1 | - | 1, 2 | 172 | |||
AK162087.1 | - | 1, 2 | 151 | |||
BC003457.1 | - | 1, 2 | 147 | 2648452 | ||
CA559663.1 | - | 1, 2 | 412 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
