siRNA Details
| Gene Symbol: Cenpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134046.5 | NP_598807.2 | 3 | 707 | ||
| GenBank mRNA | AK013198.1 | - | 3 | 379 | ||
| AK018444.1 | - | 3 | 352 | |||
| AK029226.1 | - | 3 | 708 | |||
| AK134266.1 | - | 3 | 379 | |||
| BC034399.1 | - | 3 | 366 | 4481047 | ||

Cenpo
Genomic Map